Assay Details
Gene Symbol: PPP1R3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614150.3 | XP_005614207.2 | 1030 | 92 | ||
XM_014728847.2 | XP_014584333.2 | 1030 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
