Assay Details
Gene Symbol: PPP1R3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014728847.2 | XP_014584333.2 | 1092 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
