Assay Details
Gene Symbol: AKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014728849.2 | XP_014584335.2 | 412 | 107 | ||
XM_023633357.1 | XP_023489125.1 | 449 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
