Assay Details
Gene Symbol: KLF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014728902.2 | XP_014584388.1 | 1297 | 72 | ||
XM_014728903.2 | XP_014584389.1 | 787 | 72 | |||
XM_023634474.1 | XP_023490242.1 | 728 | 72 | |||
XM_023634475.1 | XP_023490243.1 | 1000 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
