Assay Details
Gene Symbol: AWAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496730.3 | XP_001496780.1 | 497 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
