Assay Details
Gene Symbol: IGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634097.1 | XP_023489865.1 | 792 | 79 | ||
XM_023634098.1 | XP_023489866.1 | 809 | 79 | |||
XM_023634100.1 | XP_023489868.1 | 809 | 79 | |||
XM_023634101.1 | XP_023489869.1 | 679 | 79 | |||
GenBank mRNA | JL618904.1 | - | 1 - 2 | 189 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
