Assay Details
Gene Symbol: PGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634183.1 | XP_023489951.1 | 525 | 73 | ||
GenBank mRNA | JL616101.1 | - | 4 - 5 | 440 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
