Assay Details
Gene Symbol: GLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492649.4 | XP_001492699.3 | 318 | 62 | ||
XM_023634342.1 | XP_023490110.1 | 336 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
