Assay Details
Gene Symbol: PRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491375.4 | XP_001491425.1 | 279 | 72 | ||
XM_023633279.1 | XP_023489047.1 | 280 | 72 | |||
GenBank mRNA | JL616196.1 | - | 1 - 2 | 260 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
