Assay Details
Gene Symbol: SEPTIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014729135.2 | XP_014584621.1 | 863 | 92 | ||
XM_014729136.2 | XP_014584622.1 | 744 | 92 | |||
XM_023634069.1 | XP_023489837.1 | 861 | 92 | |||
XM_023634070.1 | XP_023489838.1 | 861 | 92 | |||
XM_023634071.1 | XP_023489839.1 | 861 | 92 | |||
XM_023634072.1 | XP_023489840.1 | 860 | 92 | |||
XM_023634073.1 | XP_023489841.1 | 859 | 92 | |||
XM_023634074.1 | XP_023489842.1 | 859 | 92 | |||
XM_023634075.1 | XP_023489843.1 | 857 | 92 | |||
XM_023634076.1 | XP_023489844.1 | 858 | 92 | |||
XM_023634077.1 | XP_023489845.1 | 858 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
