Assay Details
Gene Symbol: SEPTIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014729135.2 | XP_014584621.1 | 1429 | 91 | ||
XM_014729136.2 | XP_014584622.1 | 1310 | 91 | |||
XM_023634069.1 | XP_023489837.1 | 1427 | 91 | |||
XM_023634070.1 | XP_023489838.1 | 1427 | 91 | |||
XM_023634071.1 | XP_023489839.1 | 1427 | 91 | |||
XM_023634072.1 | XP_023489840.1 | 1426 | 91 | |||
XM_023634073.1 | XP_023489841.1 | 1425 | 91 | |||
XM_023634074.1 | XP_023489842.1 | 1425 | 91 | |||
XM_023634075.1 | XP_023489843.1 | 1423 | 91 | |||
XM_023634076.1 | XP_023489844.1 | 1424 | 91 | |||
XM_023634077.1 | XP_023489845.1 | 1424 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
