Assay Details
Gene Symbol: AKAP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614540.3 | XP_005614597.1 | 627 | 85 | ||
XM_014729142.2 | XP_014584628.1 | 601 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
