Assay Details
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490011.5 | XP_001490061.1 | 940 | 88 | ||
XM_005614531.3 | XP_005614588.1 | 520 | 88 | |||
GenBank mRNA | HQ889867.1 | - | 2 - 3 | 97 | 88 | |
JL616379.1 | - | 4 - 5 | 566 | 88 | ||
KX815964.1 | - | 4 - 5 | 409 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
