Assay Details
Gene Symbol: HMGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633353.1 | XP_023489121.1 | 586 | 79 | ||
XM_023633354.1 | XP_023489122.1 | 557 | 79 | |||
XM_023633355.1 | XP_023489123.1 | 567 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
