Assay Details
Gene Symbol: GAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494489.6 | XP_001494539.3 | 1592 | 63 | ||
XM_005614673.2 | XP_005614730.1 | 1597 | 63 | |||
XM_014729313.2 | XP_014584799.1 | 1595 | 63 | |||
XM_014729314.2 | XP_014584800.1 | 1899 | 63 | |||
XM_014729315.2 | XP_014584801.1 | 1789 | 63 | |||
XM_023634373.1 | XP_023490141.1 | 2275 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
