Assay Details
Gene Symbol: MTCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614660.3 | XP_005614717.1 | 5244 | 71 | ||
XM_023633328.1 | XP_023489096.1 | 517 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
