Assay Details
Gene Symbol: VBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614659.3 | XP_005614716.2 | 1280 | 65 | ||
XM_023634326.1 | XP_023490094.1 | 1280 | 65 | |||
XM_023634327.1 | XP_023490095.1 | 1287 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
