Assay Details
Gene Symbol: GPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495246.6 | XP_001495296.1 | 713 | 146 | ||
XM_023647167.1 | XP_023502935.1 | 800 | 146 | |||
XM_023647168.1 | XP_023502936.1 | 787 | 146 | |||
XM_023647169.1 | XP_023502937.1 | 704 | 146 | |||
GenBank mRNA | HQ890018.1 | - | 3 - 4 | 260 | 146 | |
JL616486.2 | - | 4 - 5 | 503 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
