Assay Details
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497639.5 | XP_001497689.3 | 672 | 105 | ||
XM_005613276.3 | XP_005613333.2 | 760 | 105 | |||
XM_014728078.2 | XP_014583564.2 | 753 | 105 | |||
XM_023648951.1 | XP_023504719.1 | 757 | 105 | |||
XM_023648952.1 | XP_023504720.1 | 1734 | 105 | |||
XM_023648953.1 | XP_023504721.1 | 2668 | 105 | |||
GenBank mRNA | HQ890259.1 | - | 4 - 5 | 497 | 105 | |
JL628166.1 | - | 2 - 3 | 149 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
