Assay Details
Gene Symbol: SMNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639623.1 | XP_023495391.1 | 560 | 67 | ||
XM_023639630.1 | XP_023495398.1 | 925 | 67 | |||
XM_023639637.1 | XP_023495405.1 | 608 | 67 | |||
GenBank mRNA | HQ890289.1 | - | 1 - 2 | 67 | 67 | |
JL616033.2 | - | 2 - 3 | 298 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
