Assay Details
Gene Symbol: DONSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630075.1 | XP_023485843.1 | 2136 | 111 | ||
GenBank mRNA | JL617664.1 | - | 1 | 327 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
