Assay Details
Gene Symbol: N4BP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621936.1 | XP_023477704.1 | 561 | 99 | ||
GenBank mRNA | JL618422.1 | - | 1 - 2 | 78 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
