Assay Details
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641298.1 | XP_023497066.1 | 1400 | 70 | ||
GenBank mRNA | JL619669.1 | - | 1 - 2 | 1360 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
