Assay Details
Gene Symbol: PELO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497587.6 | XP_001497637.1 | 3229 | 90 | ||
GenBank mRNA | JL620201.1 | - | 1 | 1023 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
