Assay Details
Gene Symbol: BCS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492073.3 | XP_001492123.1 | 391 | 82 | ||
XM_005610613.2 | XP_005610670.1 | 510 | 82 | |||
GenBank mRNA | JL622902.1 | - | 1 - 2 | 57 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
