Assay Details
Gene Symbol: CUTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014733157.2 | XP_014588643.1 | 587 | 54 | ||
GenBank mRNA | JL625988.1 | - | 2 - 3 | 230 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
