Assay Details
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023652713.1 | XP_023508481.1 | 1390 | 97 | ||
XM_023652719.1 | XP_023508487.1 | 1405 | 97 | |||
XM_023652724.1 | XP_023508492.1 | 1474 | 97 | |||
XM_023652735.1 | XP_023508503.1 | 1144 | 97 | |||
GenBank mRNA | JL626815.1 | - | 1 - 2 | 434 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
