Assay Details
Gene Symbol: ALKBH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493933.4 | XP_001493983.1 | 1664 | 104 | ||
XM_014735744.2 | XP_014591230.1 | 2034 | 104 | |||
GenBank mRNA | JL627583.1 | - | 1 | 914 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
