Assay Details
Gene Symbol: MPZL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611612.3 | XP_005611669.1 | 673 | 71 | ||
GenBank mRNA | JL628511.1 | - | 1 - 2 | 136 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
