Assay Details
Gene Symbol: NFE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639393.1 | XP_023495161.1 | 1151 | 85 | ||
GenBank mRNA | JL629974.1 | - | 1 - 2 | 517 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
