Assay Details
Gene Symbol: TMIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653174.1 | XP_023508942.1 | 699 | 81 | ||
GenBank mRNA | JL630261.1 | - | 1 - 2 | 87 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
