Assay Details
Gene Symbol: MPDU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504782.5 | XP_001504832.3 | 884 | 61 | ||
XM_023653461.1 | XP_023509229.1 | 819 | 61 | |||
GenBank mRNA | JL636981.1 | - | 1 - 2 | 346 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
