Assay Details
Gene Symbol: LTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491333.5 | XP_001491383.2 | 2936 | 126 | ||
XM_005603717.3 | XP_005603774.1 | 3059 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
