Assay Details
Gene Symbol: PCGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642787.1 | XP_023498555.1 | 6166 | 107 | ||
XM_023642796.1 | XP_023498564.1 | 6022 | 107 | |||
XM_023642803.1 | XP_023498571.1 | 6105 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
