Assay Details
Gene Symbol: MARCKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650959.1 | XP_023506727.1 | 401 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023650959.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
