Assay Details
Gene Symbol: CCDC181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609641.3 | XP_005609698.2 | 1252 | 101 | ||
XM_014739552.2 | XP_014595038.2 | 1361 | 101 | |||
XM_014739554.2 | XP_014595040.2 | 1271 | 101 | |||
XM_023640512.1 | XP_023496280.1 | 1151 | 101 | |||
XM_023640513.1 | XP_023496281.1 | 1234 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
