Assay Details
Gene Symbol: GORAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364929.4 | XP_003364977.2 | 610 | 102 | ||
XM_023640537.1 | XP_023496305.1 | 742 | 102 | |||
XM_023640538.1 | XP_023496306.1 | 468 | 102 | |||
XM_023640539.1 | XP_023496307.1 | 452 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023640539.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
