Assay Details
Gene Symbol: CENPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497104.6 | XP_001497154.1 | 1047 | 79 | ||
XM_014739547.2 | XP_014595033.2 | 513 | 79 | |||
XM_023640573.1 | XP_023496341.1 | 1117 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
