Assay Details
Gene Symbol: FAM163A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739686.2 | XP_014595172.1 | 1051 | 82 | ||
XM_023640641.1 | XP_023496409.1 | 4725 | 82 | |||
XM_023640642.1 | XP_023496410.1 | 2939 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014739686.2, XM_023640641.1, XM_023640642.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
