Assay Details
Gene Symbol: ACBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640650.1 | XP_023496418.1 | 443 | 74 | ||
XM_023640651.1 | XP_023496419.1 | 443 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
