Assay Details
Gene Symbol: KIAA1614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640654.1 | XP_023496422.1 | 3862 | 82 | ||
XM_023640655.1 | XP_023496423.1 | 3835 | 82 | |||
XM_023640656.1 | XP_023496424.1 | 3805 | 82 | |||
XM_023640657.1 | XP_023496425.1 | 4929 | 82 | |||
XM_023640658.1 | XP_023496426.1 | 4016 | 82 | |||
XM_023640660.1 | XP_023496428.1 | 3712 | 82 | |||
XM_023640661.1 | XP_023496429.1 | 3865 | 82 | |||
XM_023640664.1 | XP_023496432.1 | 5993 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
