Assay Details
Gene Symbol: STX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488916.6 | XP_001488966.2 | 741 | 82 | ||
XM_023640665.1 | XP_023496433.1 | 741 | 82 | |||
XM_023640666.1 | XP_023496434.1 | 741 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
