Assay Details
Gene Symbol: GLUL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489235.6 | XP_001489285.4 | 754 | 75 | ||
XM_023640677.1 | XP_023496445.1 | 755 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
