Assay Details
Gene Symbol: RGS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489479.4 | XP_001489529.1 | 340 | 74 | ||
XM_005609765.3 | XP_005609822.1 | 337 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
