Assay Details
Gene Symbol: RNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640704.1 | XP_023496472.1 | 965 | 81 | ||
GenBank mRNA | JL621089.1 | - | 2 - 3 | 390 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
