Assay Details
Gene Symbol: VASH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364952.4 | XP_003365000.2 | 1403 | 83 | ||
XM_014739642.2 | XP_014595128.2 | 1359 | 83 | |||
XM_023640745.1 | XP_023496513.1 | 1603 | 83 | |||
XM_023640746.1 | XP_023496514.1 | 1272 | 83 | |||
XM_023640747.1 | XP_023496515.1 | 918 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
