Assay Details
Gene Symbol: FLVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488684.6 | XP_001488734.2 | 942 | 72 | ||
XM_005609805.3 | XP_005609862.1 | 943 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
