Assay Details
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618890.1 | XP_023474658.1 | 383 | 72 | ||
XM_023618891.1 | XP_023474659.1 | 382 | 72 | |||
XM_023618893.1 | XP_023474661.1 | 377 | 72 | |||
XM_023618894.1 | XP_023474662.1 | 379 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
