Assay Details
Gene Symbol: LOC111768129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619299.1 | XP_023475067.1 | 777 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023619299.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
