Assay Details
Gene Symbol: C2CD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023622363.1 | XP_023478131.1 | 5418 | 83 | ||
XM_023622364.1 | XP_023478132.1 | 5415 | 83 | |||
XM_023622365.1 | XP_023478133.1 | 5401 | 83 | |||
XM_023622366.1 | XP_023478134.1 | 5219 | 83 | |||
XM_023622367.1 | XP_023478135.1 | 5381 | 83 | |||
XM_023622368.1 | XP_023478136.1 | 5245 | 83 | |||
XM_023622369.1 | XP_023478137.1 | 5343 | 83 | |||
XM_023622370.1 | XP_023478138.1 | 5383 | 83 | |||
XM_023622371.1 | XP_023478139.1 | 5256 | 83 | |||
XM_023622372.1 | XP_023478140.1 | 5238 | 83 | |||
XM_023622373.1 | XP_023478141.1 | 5125 | 83 | |||
XM_023622374.1 | XP_023478142.1 | 5121 | 83 | |||
XM_023622375.1 | XP_023478143.1 | 5183 | 83 | |||
XM_023622376.1 | XP_023478144.1 | 5048 | 83 | |||
XM_023622377.1 | XP_023478145.1 | 5146 | 83 | |||
XM_023622378.1 | XP_023478146.1 | 5058 | 83 | |||
XM_023622379.1 | XP_023478147.1 | 5042 | 83 | |||
XM_023622380.1 | XP_023478148.1 | 4925 | 83 | |||
XM_023622381.1 | XP_023478149.1 | 4879 | 83 | |||
XM_023622382.1 | XP_023478150.1 | 1881 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
